Sartorius Arium® Advance EDI Type 2 Water Purification Systems
Compared with conventional water systems, the Arium® Advance EDI with its iJust software makes the best use of your initial feed water conditions and water resources
  • Consistently high Type 2 water quality thanks to the latest EDI technology
  • Optimized, lean and green water usage
  • Use of innovative Bag technology, eliminates costly tank cleaning
  • Display with touch function and intuitive menu is easy to use
The Arium® Advance EDI consistently delivers Type 2 water of the best quality at rates of up to 5 or 10 L/h. This system makes the best use of your valuable water resources by automatically optimizing water usage with its intelligent I-Just software. Moreover, the Arium® Advance EDI attains the highest retention rates of ions, while providing an optimum yield of purified water. As a result, the Arium® Advance EDI reliably removes oxidants, heavy metal ions and particulates from feed water. In addition to prefiltration and purification by reverse osmosis modules, electrochemical deionization is carried out as a third purification step. By means of this modern EDI technology, the Arium® Advance EDI reliably guarantees a high effective removal of ions contained in the feed water. Pure water produced by Arium®Advance EDI is stored in the unique, closed Arium®Bagtank system, which reliably protects your purified water from secondary contamination. Time-consuming tank cleaning intervals are eliminated thanks to the interchangeable bag. This ensures that you obtain the best water quality and, therefore, reproducible results.
Intelligent iJust enhances the quality of purified water, reduces water usage, and calculates the optimal cleaning cycles for the system. iJust stands for innovative technology that optimizes water production. Amongst other things, the intelligent Arium® software controls a valve on the concentrate outlet in accordance with the data that have been entered for CaCO3 and CO2. iJust therefore optimizes the product water quality and water consumption.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description ASTM Water Grade Power (VAC)
Availability Pricing
Sartorius Arium® Advance EDI Wall-Mounted System, Flow Capacity 5 L/h 9929342 Mfr # H2O-EDI-1-B
COMPARE
Arium® Advance EDI Wall-Mounted System, Flow Capacity 5 L/h Type 2 100 to 240
Call For Price
Sartorius Arium® Advance EDI Benchtop System, Flow Capacity 5 L/h 9929343 Mfr # H2O-EDI-1-T
COMPARE
Arium® Advance EDI Benchtop System, Flow Capacity 5 L/h Type 2 100 to 240
Call For Price
Sartorius Arium® Advance EDI Wall-Mounted System, Flow Capacity 10 L/h 9929344 Mfr # H2O-EDI-2-B
COMPARE
Arium® Advance EDI Wall-Mounted System, Flow Capacity 10 L/h Type 2 100 to 240
Call For Price
Sartorius Arium® Advance EDI Benchtop System, Flow Capacity 10 L/h 9929345 Mfr # H2O-EDI-2-T
COMPARE
Arium® Advance EDI Benchtop System, Flow Capacity 10 L/h Type 2 100 to 240