SaniSure® Cap2V8® Aseptic Centrifuge Tube Assemblies, Falcon® Tubes
Reduce contamination and product loss with one-piece molded tube and cap assemblies
  • Ideal for aseptic transfer, storage and sampling
  • Cost-effective—reduces assembly and installation time
  • Lot-traceable
  • Features two ports (vent and transfer) with 1/8"ID x 1/4"OD tubing
  • Meets USP Class VI requirements
The single-use centrifuge assemblies are ideal for aseptic storage, transfer, and sampling. One continuous flow path reduces contamination chances and product loss. Each assembly includes a centrifuge tube and molded cap with transfer and vent tubing.
17,500.00 - ₹23,200.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features Sterile Tube Capacity (mL)
Availability Pricing
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly with Dip Tube (Falcon®), TPE Tubing, 15 mL, Sterile 5014277 Mfr # CAP2V8-15C-A-Y-GIDIP
COMPARE
Dip tube, 8"L transfer line, and 2"L vent line (TPE tubing) Yes 15
₹18,100.00INR / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly with Dip Tube (Falcon®), TPE Tubing, 15 mL, Nonsterile 5014278 Mfr # CAP2V8-15C-A-Y-NGDI
COMPARE
Dip tube, 8"L transfer line, and 2"L vent line (TPE tubing) No 15
₹17,500.00INR / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly (Falcon®), PVC Tubing, 50 mL, Sterile 5014279 Mfr # CAP2V8 -50C - A - C -GI
COMPARE
12"L transfer line and 4"L vent line (PVC tubing) Yes 50
₹22,600.00INR / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly with Dip Tube (Falcon®), PVC Tubing, 50 mL, Sterile 5014280 Mfr # CAP2V8 -50C - A - C -GI - DIP
COMPARE
Dip tube, 12"L transfer line, and 4"L vent line (PVC tubing) Yes 50
₹23,200.00INR / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly (Falcon®), PVC Tubing, 50 mL, Nonterile 5014281 Mfr # CAP2V8 -50C - A - C -NG
COMPARE
12"L transfer line and 4"L vent line (PVC tubing) No 50
₹21,600.00INR / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly with Dip Tube (Falcon®), PVC Tubing, 50 mL, Nonterile 5014282 Mfr # CAP2V8 -50C - A - C -NG - DIP
COMPARE
Dip tube, 12"L transfer line, and 4"L vent line (PVC tubing) No 50
₹22,200.00INR / Each