Sampling Tubes
Superior blade design offers easy hand operated penetration, even in hard soils
  • Nonremovable blade stays sharp
  • There's no need to bother with stripped threads typical of removable tips
Sharp blade produces smooth, well-formed core. Choose from dry or wet/dry sampling tubes. Tubes are made of easy to clean nickel-plated carbon steel.
29,300.00 - ₹62,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Core Diameter (cm) Length (cm) Material
Availability Pricing
COMPARE
1.91 30.48 Nickel-plated carbon steel
₹29,300.00INR / Each
COMPARE
3.18 30.48 Nickel-plated carbon steel
₹58,900.00INR / Each
COMPARE
3.18 45.72 Nickel-plated carbon steel
₹62,200.00INR / Each