Sample Bottle Carrying Rack
Use this vinyl coated rack in the laboratory to conveniently transport bottles
  • Rack features a double grid design
  • Autoclavable
Use rack with sample bottles 06043-01, 06049-01, 06050-01, and 06020-64 on this page, or with any bottle with a maximum outer diameter of 2.5". Rack measures 13.5"L x 11"W x 4"H; the 20 compartments measure 2.5 x 2.5".
22,800.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material Overall Height (in)
Availability Pricing
Nasco Sampling B01014 Carrying Rack for 20, 18 oz Whirl-Pak Bags or 2.5" Diameter Bottles 6620300 Mfr # B01014
COMPARE
Vinyl 4
₹22,800.00INR / Each