Salisbury by Honeywell Insulated Clampsticks
Clampsticks help increase your distance from live circuits. Used for attaching and detaching grounding connectors and other clamps during electrical work.
* Constructed of closed cell foam-filled tubular fiberglass in accordance with ASTM F711
* Tough thermoplastic head ferrule
* Wide 15/16" (238 mm) opening
* Heavy rubber end cap
* Easy grip handle
* Clampsticks meet ASTM F1825
380,000.00 - ₹504,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Diameter (InchShort) Length (InchShort)
Availability Pricing
Honeywell 4007 Salisbury External Rod Clampsticks, 4 foot 8" length 8643703 Mfr # 4007-ES
COMPARE
3/8 56
₹406,000.00INR / Each
Honeywell 4008 Salisbury External Rod Clampsticks, 5 foot 8" length 8643704 Mfr # 4008
COMPARE
3/8 68
₹420,000.00INR / Each
Honeywell 4009 Salisbury External Rod Clampsticks, 6 foot 8" length 8643705 Mfr # 4009
COMPARE
3/8 80
₹380,000.00INR / Each
Honeywell 4010 Salisbury External Rod Clampsticks, 8 foot 8" length 8643706 Mfr # 4010-ES
COMPARE
3/8 104
₹504,000.00INR / Each