Saint-Gobain Tygopure® Monobarb Sanitary Fittings
Engineered for a smooth, secure, and contamination-free connection
  • Smooth inner surface minimizes particle entrapment and bacterial growth
  • Leak-proof security ensures reliable tubing connections
  • Tapered design reduces turbulence and stagnation
  • Available in three radiation-sterilizable polymer materials
  • USP Class VI compliant to ensure the highest biopharmaceutical safety standards
Tygopure® Monobarb Sanitary Fittings are designed for use in bioreactors, filtration systems, media mixing containers, and other critical biopharmaceutical processes. The innovative one-piece monobarb design prevents leaks, reduces contamination risks, and provides a seamless fluid path for gentle and efficient fluid transfer. Available in polypropylene, PVDF, and polysulfone, these fittings are radiation-sterilizable, offering a complete solution for secure and sterile fluid management.
7,700.00 - ₹12,100.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material
Availability Pricing
Saint Gobain Tygopure® Sanitary Coupling, 1/4", Polypropylene; 1/Each 5013450 Mfr # ATP250PPSB
COMPARE
1/4 Polypropylene
₹7,700.00INR / Each
Saint Gobain Tygopure® Sanitary Coupling, 3/8", Polypropylene; 1/Each 5013451 Mfr # ATP375PPSB
COMPARE
3/8 Polypropylene
₹7,700.00INR / Each
Saint Gobain Tygopure® Sanitary Coupling, 1/2", Polypropylene; 1/Each 5013452 Mfr # ATP500PPSB
COMPARE
1/2 Polypropylene
₹7,900.00INR / Each
Saint Gobain Tygopure® Sanitary Coupling, 1/4", PVDF; 1/Each 5013453 Mfr # ATP250PFSB
COMPARE
1/4 PVDF
₹10,000.00INR / Each
Saint Gobain Tygopure® Sanitary Coupling, 3/8", PVDF; 1/Each 5013454 Mfr # ATP375PFSB
COMPARE
3/8 PVDF
₹10,400.00INR / Each
Saint Gobain Tygopure® Sanitary Coupling, 1/2", PVDF; 1/Each 5013455 Mfr # ATP500PFSB
COMPARE
1/2 PVDF
₹10,800.00INR / Each
Saint Gobain Tygopure® Sanitary Coupling, 3/8", Polysulfone; 1/Each 5013457 Mfr # ATP375PSSB
COMPARE
3/8 Polysulfone
₹11,100.00INR / Each
Saint Gobain Tygopure® Sanitary Coupling, 1/2", Polysulfone; 1/Each 5013458 Mfr # ATP500PSSB
COMPARE
1/2 Polysulfone
₹12,100.00INR / Each
Showing 1- 8 of 8