Saint-Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks
Multilayer liners provide outstanding bio and chemical compatibility, structural strength, and gas barrier—excellent for many demanding bioprocess applications
  • Easily integrate into your single-use process applications
  • Minimize the risk of cross-contamination and reduce cleaning time
  • Place inside tank or container—open-top bag accommodates a topdown paddle mixer
  • Bags have 5 mil (side wall) and 7 mil (bottom) film thickness
  • Free of any animal-derived components for greater process purity
  • Manufactured and packed in an ISO Class 7 certified cleanroom
  • QSR and ISO 9001:2015 compliant
  • Meet the requirements of USP Class VI and ISO 10993-3 testing
These single-use bioprocess bags feature multilayer films in different materials to provide the best biocompatibility, chemical resistance, and gas barrier to meet your bioprocessing applications. The LLDPE fluid contact layer offers outstanding biocompatibility with broad chemical compatibility for safe handling of a wide range of biopharmaceutical liquids. The EVOH middle layer ensures minimum gas permeation, ensuring the integrity of the fluid. The impact and tear resistant outside nylon layer offers outstanding durability. A multi-material tie-structure bonds the three layers, adding to the overall performance of the bag. Bags are ideal for use in buffer and media storage, bioreactor feed and harvest, and chromatography feed.
Place a liner inside a tank or containment vessel. This open-top bag accommodates a topdown paddle mixer, commonly used in upstream media preparation. Use bags in temperatures ranging from –80 to 60°C (–112 to 140°F). Bags are sterilized by gamma irradiation, ports are capped and covered, and systems are double bagged for integrity.
180,000.00 - ₹312,000.00INR / Pkg of 20
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters)
Availability Pricing
Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 20 L; 20/Pk 4852405 Mfr # LR0020L
COMPARE
20
₹264,000.00INR / Pkg of 20
Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 50 L; 20/Pk 4852406 Mfr # LR0050L
COMPARE
50
₹284,000.00INR / Pkg of 20
Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 100 L; 20/Pk 4852407 Mfr # LR0100L
COMPARE
100
₹312,000.00INR / Pkg of 20
Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 200 L; 10/Pk 4852408 Mfr # LR0200L
COMPARE
200
₹180,000.00INR / Pkg of 10
Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 300 L; 10/Pk 4852409 Mfr # LR0300L
COMPARE
300
₹207,000.00INR / Pkg of 10
Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 400 L; 10/Pk 4852410 Mfr # LR0400L
COMPARE
400
₹245,000.00INR / Pkg of 10
Saint Gobain Single-Use 3D Bioprocess Liners for Cylindrical Tanks, 500 L; 10/Pk 4852411 Mfr # LR0500L
COMPARE
500
₹287,000.00INR / Pkg of 10
Showing 1- 7 of 7