Saint-Gobain Single-Use 3D Bioprocess Bags for Rectangular Tanks with 4 Ports, 30"L Tubing, and Bottom Drain
Multilayer bags provide outstanding bio and chemical compatibility, structural strength, and gas barrier—excellent for many demanding bioprocess applications
  • Easily integrate into your single-use process applications
  • Minimize the risk of cross-contamination and reduce cleaning time
  • 3D bags fit perfectly in rectangular tanks
  • Three top ports with C-Flex® 374 tubing and fittings, including a needle-free sampling port, plus a drain port
  • Free of any animal-derived components for greater process purity
  • Manufactured and packed in an ISO Class 7 certified cleanroom
  • QSR and ISO 9001:2015 compliant
  • Meet the requirements of USP Class VI and ISO 10993-3 testing
These single-use bioprocess bags feature multilayer films in different materials to provide the best biocompatibility, chemical resistance, and gas barrier to meet your bioprocessing applications. The LLDPE fluid contact layer offers outstanding biocompatibility with broad chemical compatibility for safe handling of a wide range of biopharmaceutical liquids. The EVOH middle layer ensures minimum gas permeation, ensuring the integrity of the fluid. The impact and tear resistant outside nylon layer offers outstanding durability. A multi-material tie-structure bonds the three layers, adding to the overall performance of the bag. Bags are ideal for use in buffer and media storage, bioreactor feed and harvest, and chromatography feed.
A 3D bag consists of multilayer sheets sealed together to often form a cuboidal shape and fits inside a supporting tank or container. These collapsible bags minimize the risk of cross-contamination and reduce cleaning and turnaround time. Each bag features four ports with 30” long clear C-Flex® 374 tubing, MPX inserts (on three ports) or female locking Luer for needle-free sampling, and pinch clamps. Use bags in temperatures ranging from –80 to 60°C (–112 to 140°F). Bags are sterilized by gamma irradiation, ports are capped and covered, and systems are double bagged for integrity.
356,000.00 - ₹415,000.00INR / Pkg of 4
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters)
Availability Pricing
Saint Gobain Single-Use 3D Bioprocess Bags for Rectangular Tanks, 200 L, 4 Ports, 30"L Tubing, Bottom Drain; 4/Pk 4852420 Mfr # BP0200LB
COMPARE
200
₹356,000.00INR / Pkg of 4
Saint Gobain Single-Use 3D Bioprocess Bags for Rectangular Tanks, 500 L, 4 Ports, 30"L Tubing, Bottom Drain; 4/Pk 4852421 Mfr # BP0500LB
COMPARE
500
₹415,000.00INR / Pkg of 4