Saint-Gobain Single-Use 2D Bioprocess Bags with 3 Ports and 6"L Tubing
Multilayer bags provide outstanding bio and chemical compatibility, structural strength, and gas barrier—excellent for many demanding bioprocess applications
  • Easily integrate into your single-use process applications
  • Minimize the risk of cross-contamination and reduce cleaning time
  • 2D bags are made by welding two multilayer sheets together
  • Three ports with clear C-Flex® 374 tubing and fittings, including a needle-free sampling port
  • Free of any animal-derived components for greater process purity
  • Manufactured and packed in an ISO Class 7 certified cleanroom
  • QSR and ISO 9001:2015 compliant
  • Meet the requirements of USP Class VI and ISO 10993-3 testing
These single-use bioprocess bags feature multilayer films in different materials to provide the best biocompatibility, chemical resistance, and gas barrier to meet your bioprocessing applications. The LLDPE fluid contact layer offers outstanding biocompatibility with broad chemical compatibility for safe handling of a wide range of biopharmaceutical liquids. The EVOH middle layer ensures minimum gas permeation, ensuring the integrity of the fluid. The impact and tear resistant outside nylon layer offers outstanding durability. A multi-material tie-structure bonds the three layers, adding to the overall performance of the bag. Bags are ideal for use in buffer and media storage, bioreactor feed and harvest, and chromatography feed.
The 2D bags are manufactured by welding two multilayer sheets together. These collapsible bags minimize the risk of cross-contamination and reduce cleaning and turnaround time. Place bags flat in rocker bioreactor systems or hang using an insert hole at the top of the bag. Each bag features three ports with 6” long clear C-Flex® 374 tubing, MPC inserts (on two ports) or female locking Luer with needle-free sampling port, and pinch clamps. Use bags in temperatures ranging from –80 to 60°C (–112 to 140°F). Bags are sterilized by gamma irradiation, ports are capped and covered, and systems are double bagged for integrity.
157,000.00 - ₹290,000.00INR / Pkg of 20
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL)
Availability Pricing
Saint Gobain Single-Use 2D Bioprocess Bags, 50 mL, 3 Ports, 6"L Tubing; 20/Pk 4852386 Mfr # BP0050M
COMPARE
50
₹200,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 150 mL, 3 Ports, 6"L Tubing; 20/Pk 4852387 Mfr # BP0150M
COMPARE
150
₹218,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 250 mL, 3 Ports, 6"L Tubing; 20/Pk 4852388 Mfr # BP0250M
COMPARE
250
₹229,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 500 mL, 3 Ports, 6"L Tubing; 20/Pk 4852389 Mfr # BP0500M
COMPARE
500
₹232,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 1 L, 3 Ports, 6"L Tubing; 20/Pk 4852390 Mfr # BP0001L
COMPARE
1000
₹254,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 2 L, 3 Ports, 6"L Tubing; 20/Pk 4852391 Mfr # BP0002L
COMPARE
2000
₹271,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 3 L, 3 Ports, 6"L Tubing; 20/Pk 4852392 Mfr # BP0003L
COMPARE
3000
₹273,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 5 L, 3 Ports, 6"L Tubing; 20/Pk 4852393 Mfr # BP0005L
COMPARE
5000
₹290,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 10 L, 3 Ports, 6"L Tubing; 10/Pk 4852394 Mfr # BP0010L
COMPARE
10000
₹157,000.00INR / Pkg of 10
Saint Gobain Single-Use 2D Bioprocess Bags, 20 L, 3 Ports, 6"L Tubing; 10/Pk 4852395 Mfr # BP0020L
COMPARE
20000
₹169,000.00INR / Pkg of 10
Showing 1- 10 of 10