Saint-Gobain Single-Use 2D Bioprocess Bags with 3 Ports and 20"L Tubing
Multilayer bags provide outstanding bio and chemical compatibility, structural strength, and gas barrier—excellent for many demanding bioprocess applications
  • Easily integrate into your single-use process applications
  • Minimize the risk of cross-contamination and reduce cleaning time
  • 2D bags are made by welding two multilayer sheets together
  • Three ports with clear C-Flex® 374 tubing and fittings, including a needle-free sampling port
  • Free of any animal-derived components for greater process purity
  • Manufactured and packed in an ISO Class 7 certified cleanroom
  • QSR and ISO 9001:2015 compliant
  • Meet the requirements of USP Class VI and ISO 10993-3 testing
These single-use bioprocess bags feature multilayer films in different materials to provide the best biocompatibility, chemical resistance, and gas barrier to meet your bioprocessing applications. The LLDPE fluid contact layer offers outstanding biocompatibility with broad chemical compatibility for safe handling of a wide range of biopharmaceutical liquids. The EVOH middle layer ensures minimum gas permeation, ensuring the integrity of the fluid. The impact and tear resistant outside nylon layer offers outstanding durability. A multi-material tie-structure bonds the three layers, adding to the overall performance of the bag. Bags are ideal for use in buffer and media storage, bioreactor feed and harvest, and chromatography feed.
The 2D bags are manufactured by welding two multilayer sheets together. These collapsible bags minimize the risk of cross-contamination and reduce cleaning and turnaround time. Place bags flat in rocker bioreactor systems or hang using an insert hole at the top of the bag. Each bag features three ports with 20” long clear C-Flex® 374 tubing, MPC inserts (on two ports) or female locking Luer with needle-free sampling port, and pinch clamps. Use bags in temperatures ranging from –80 to 60°C (–112 to 140°F). Bags are sterilized by gamma irradiation, ports are capped and covered, and systems are double bagged for integrity.
179,000.00 - ₹331,000.00INR / Pkg of 20
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL)
Availability Pricing
Saint Gobain Single-Use 2D Bioprocess Bags, 150 mL, 3 Ports, 20"L Tubing; 20/Pk 4852396 Mfr # BP0150ML
COMPARE
150
₹240,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 250 mL, 3 Ports, 20"L Tubing; 20/Pk 4852397 Mfr # BP0250ML
COMPARE
250
₹252,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 500 mL, 3 Ports, 20"L Tubing; 20/Pk 4852398 Mfr # BP0500ML
COMPARE
500
₹264,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 1 L, 3 Ports, 20"L Tubing; 20/Pk 4852399 Mfr # BP0001LL
COMPARE
1000
₹278,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 2 L, 3 Ports, 20"L Tubing; 20/Pk 4852400 Mfr # BP0002LL
COMPARE
2000
₹294,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 3 L, 3 Ports, 20"L Tubing; 20/Pk 4852401 Mfr # BP0003LL
COMPARE
3000
₹312,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 5 L, 3 Ports, 20"L Tubing; 20/Pk 4852402 Mfr # BP0005LL
COMPARE
5000
₹331,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 10 L, 3 Ports, 20"L Tubing; 10/Pk 4852403 Mfr # BP0010LL
COMPARE
10000
₹179,000.00INR / Pkg of 10
Saint Gobain Single-Use 2D Bioprocess Bags, 20 L, 3 Ports, 20"L Tubing; 10/Pk 4852404 Mfr # BP0020LL
COMPARE
20000
₹193,000.00INR / Pkg of 10
Showing 1- 9 of 9