Saint-Gobain Single-Use 2D Bioprocess Bags with 2 Ports and 12"L Tubing
Multilayer bags provide outstanding bio and chemical compatibility, structural strength, and gas barrier—excellent for many demanding bioprocess applications
  • Easily integrate into your single-use process applications
  • Minimize the risk of cross-contamination and reduce cleaning time
  • 2D bags are made by welding two multilayer sheets together
  • Two ports with clear C-Flex® 374 tubing and fittings, including a needle-free sampling port
  • Free of any animal-derived components for greater process purity
  • Manufactured and packed in an ISO Class 7 certified cleanroom
  • QSR and ISO 9001:2015 compliant
  • Meet the requirements of USP Class VI and ISO 10993-3 testing
These single-use bioprocess bags feature multilayer films in different materials to provide the best biocompatibility, chemical resistance, and gas barrier to meet your bioprocessing applications. The LLDPE fluid contact layer offers outstanding biocompatibility with broad chemical compatibility for safe handling of a wide range of biopharmaceutical liquids. The EVOH middle layer ensures minimum gas permeation, ensuring the integrity of the fluid. The impact and tear resistant outside nylon layer offers outstanding durability. A multi-material tie-structure bonds the three layers, adding to the overall performance of the bag. Bags are ideal for use in buffer and media storage, bioreactor feed and harvest, and chromatography feed.
The 2D bags are manufactured by welding two multilayer sheets together. These collapsible bags minimize the risk of cross-contamination and reduce cleaning and turnaround time. Place bags flat in rocker bioreactor systems or hang using an insert hole at the top of the bag. Each bag features two ports with 12” long clear C-Flex® 374 tubing, male locking Luer, female locking Luer with needle-free sampling port, and pinch clamps. Use bags in temperatures ranging from –80 to 60°C (–112 to 140°F). Bags are sterilized by gamma irradiation, ports are capped and covered, and systems are double bagged for integrity.
179,000.00 - ₹207,000.00INR / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL)
Availability Pricing
Saint Gobain Single-Use 2D Bioprocess Bags, 50 mL, 2 Ports, 12"L Tubing; 20/Pk 4852382 Mfr # BP0050MS
COMPARE
50
₹179,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 150 mL, 2 Ports, 12"L Tubing; 20/Pk 4852383 Mfr # BP0150MS
COMPARE
150
₹195,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 250 mL, 2 Ports, 12"L Tubing; 20/Pk 4852384 Mfr # BP0250MS
COMPARE
200
₹197,000.00INR / Pkg of 20
Saint Gobain Single-Use 2D Bioprocess Bags, 500 mL, 2 Ports, 12"L Tubing; 20/Pk 4852385 Mfr # BP0500MS
COMPARE
500
₹207,000.00INR / Pkg of 20