Saint-Gobain Pure-Fit® SIB® Hose Barb Fittings, 90° Elbow, Polypropylene
Smooth inner bore ensures seamless fluid transition between tubing and fittings
  • Unique internal design eliminates dead space
  • No areas of entrapment—prohibiting a potential for bacterial growth
  • Use with Pure-Fit and BarbLock® retainers for leak-free connections
  • Easily change the flow direction with a 90° elbow
  • Steriliza by gamma irradiation
  • Animal-derived component free and BPA free
  • Manufactured and packaged in Class 8 cleanroom
  • USP <88> Class VI and USP <87> compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Eliminate dead space and entrapment, a potential breeding ground for bacterial growth, with smooth inner bore (SIB) fittings. SIB fittings provide a seamless, “zero” transition between tubing and fittings, maintaining complete fluid integrity. Use with Pure-Fit or BarbLock retainer assembly that provides 360° compression to ensure a leak-free connection.
34,100.00 - ₹37,700.00INR / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in)
Availability Pricing
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Elbow Union, Polypropylene, 1/4"; 25/Pk 5014324 Mfr # PFE250PPAF
COMPARE
1/4
₹34,100.00INR / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Elbow Union, Polypropylene, 3/8"; 25/Pk 5014336 Mfr # PFE375PPAF
COMPARE
3/8
₹35,900.00INR / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Elbow Union, Polypropylene, 1/2"; 25/Pk 5014344 Mfr # PFE500PPAF
COMPARE
1/2
₹37,700.00INR / Pkg of 25