Saint-Gobain Bio-Simplex® Media Bottle Assembly Systems
Aseptic, single-use solutions for secure fluid handling
  • Closed-system design minimizes contamination risks
  • Low gas permeability ensures sample integrity
  • Clarity for easy content viewing
  • Volume-graduated design for precise measurements
  • Compatible with sterile tubing welding and sealing
Designed for the bioprocess industry, Bio-Simplex® Media Bottle Assembly Systems provide a reliable, closed-system solution for aseptic sample handling, fluid transfer, and storage. Featuring a two-port EZ Top® container closure made from C-Flex® biopharmaceutical thermoplastic elastomer (TPE), these bottles ensure a seamless and sterile fluid path. The 0.2 µm vent filter supports sterile gas exchange, while the thermal bond between stopper and tubing eliminates contamination risks. Available in sizes ranging from 60 mL to 2 L, these assemblies meet a Sterility Assurance Level (SAL) of 10?6, making them ideal for applications such as media storage, buffer preparation, and aseptic sampling.
21,400.00 - ₹36,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
Bio-Simplex Media Bottle Assembly Systems, 60mL 5014184 Mfr # SA24MM-60-2F
COMPARE
60 Polyethylene Terephthalate Glycol Media Bottle Assembly System, 2000 mL, 5/Pk
₹21,400.00INR / Each
Bio-Simplex Media Bottle Assembly Systems, 125mL 5014185 Mfr # SA38MM-125-2F-DB
COMPARE
125 Polyethylene Terephthalate Glycol Media Bottle Assembly System, 2000 mL, 5/Pk
₹22,900.00INR / Each
Bio-Simplex Media Bottle Assembly Systems, 250mL 5014186 Mfr # SA38MM-250-2F-DB
COMPARE
250 Polyethylene Terephthalate Glycol Media Bottle Assembly System, 2000 mL, 5/Pk
₹22,900.00INR / Each
Bio-Simplex Media Bottle Assembly Systems, 500mL 5014187 Mfr # SA38MM-500-2F-DB
COMPARE
500 Polyethylene Terephthalate Glycol Media Bottle Assembly System, 2000 mL, 5/Pk
₹23,600.00INR / Each
Bio-Simplex Media Bottle Assembly Systems, 1000mL 5014188 Mfr # SA38MM-1L-2F-DB
COMPARE
1000 Polyethylene Terephthalate Glycol Media Bottle Assembly System, 2000 mL, 5/Pk
₹26,600.00INR / Each
Bio-Simplex Media Bottle Assembly Systems,2000mL 5014189 Mfr # SA53B-2F-DB
COMPARE
2000 Polyethylene Terephthalate Glycol Media Bottle Assembly System, 2000 mL, 5/Pk
₹36,900.00INR / Each