Rotor-Stator Generators for IKA T50 ULTRA-TURRAX Heavy-Duty Homogenizer
Choose a generator that matches your sample volume and required level of dispersion
All rotor-stator generators are shielded in 316 stainless steel; interior components are made of PTFE.
405,888.00 - ₹468,384.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Overall Length (mm) Shaft Diameter (inches)
Availability Pricing
IKA 8003900 T50 ULTRA-TURRAX Rotor-Stator Generator, 0.5 to 10 L, PTFE and 316SS 0473860 Mfr # 0008003900
COMPARE
10000 304.8 1.5
₹468,384.00INR / Each
IKA 8003000 T50 ULTRA-TURRAX Rotor-Stator Generator, 0.5 to 20 L, PTFE and 316SS 0473872 Mfr # 0008003000
COMPARE
20000 304.8 1.5
₹405,888.00INR / Each
IKA 8003300 T50 ULTRA-TURRAX Rotor-Stator Generator, 0.5 to 15 L, PTFE and 316SS 0473876 Mfr # 0008003300
COMPARE
15000 304.8 1.5
₹446,320.00INR / Each