Reichert® Coolant/Battery Acid Refractometers
Monitor quality of industrial fluids and coolants with ease
  • Automatic temperature compensation provides accurate readings at any temperature
  • Large, easy-to-read scales and depth of field
  • Portable – no batteries or power cord required
  • Rugged, chemical-resistant housing
Coolant/battery acid refractometers offer an accurate, fast, and easy-to-use method for testing engine coolant freeze point, glycol concentration, and battery condition. Housing is shock-resistant, water-resistant, and dustproof. Determine the percent concentration from the freezing point reading and the included chart. Include a dipstick and flexible sampling pump built into the housing. Housing has an IP67 waterproof rating.
65,100.00 - ₹71,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Resolution
Availability Pricing
Reichert 137584L0 Coolant/Battery Acid Refractometer, -60 to 32F, 1.10 to 1.40 SG 8100000 Mfr # 137584L0
COMPARE
1 degree F 0.01 s.g.
₹71,600.00INR / Each
Reichert 137564L0 Coolant/Battery Acid Refractometer, -49 to 0C, 1.10 to 1.40 SG 8100005 Mfr # 137564L0
COMPARE
1 degree F 0.01 s.g.
₹65,100.00INR / Each