Refractive Index Standard Solutions
Calibrate your refractometer with these standard solution sets
  • All solutions are standardized at 25°C
Each set includes solutions in 0.25 oz (7.4 mL) glass bottles, plastic screw cap, and glass applicator rod.
127,000.00 - ₹383,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Refractive Index Interval Description
Availability Pricing
Cargille 18031 Refractive index standard solution; 1.300 to 1.395, 20 liquids WW-81006-00 Mfr # 18031
COMPARE
±0.0005 (RI) 0.005 Refractive index standard solution; 1.300 to 1.395, 20 liquids
₹244,000.00INR / Each
Cargille 18032 Refractive index standard solution; 1.300 to 1.390, 10 liquids WW-81006-02 Mfr # 18032
COMPARE
±0.0005 (RI) 0.01 Refractive index standard solution; 1.300 to 1.390, 10 liquids
₹132,000.00INR / Each
Cargille 18061 Refractive Index Standard Solution, 1.400 to 1.458; 7.4 mL x 30 Liquids WW-81006-04 Mfr # 18061
COMPARE
±0.0002 (RI) 0.002 Refractive index standard solution; 1.400 to 1.458, 30 liquids
₹233,000.00INR / Each
Cargille 18062 Refractive Index Standard Solution, 1.400 to 1.456; 7.4 mL x 15 liquids WW-81006-06 Mfr # 18062
COMPARE
±0.0002 (RI) 0.004 Refractive index standard solution; 1.400 to 1.456, 15 liquids
₹127,000.00INR / Each
Cargille 18092 Refractive Index Standard Solution, 1.460 to 1.640; 7.4 mL x 46 liquids WW-81006-08 Mfr # 18092
COMPARE
±0.0002 (RI) 0.004 Refractive index standard solution; 1.460 to 1.640, 46 liquids
₹383,000.00INR / Each
Cargille 18122 Refractive Index Standard Solution, 1.644 to 1.700; 7.4 mL x 15 liquids WW-81006-10 Mfr # 18122
COMPARE
±0.0002 (RI) 0.004 Refractive index standard solution; 1.644 to 1.700, 15 liquids
₹271,000.00INR / Each
Cargille 18152 Refractive Index Standard Solution, 1.710 to 1.800; 7.4 mL x 10 liquids WW-81006-14 Mfr # 18152
COMPARE
±0.0005 (RI) 0.01 Refractive index standard solution; 1.710 to 1.800, 10 liquids
₹180,000.00INR / Each
Showing 1- 7 of 7