Raven Environmental Products Coretaker Sludge Samplers
Ideal for direct measurement of sludge blanket thickness by core sample extraction
  • Lifetime, no breakage gaurantee
  • Floats for easy retrieval
  • No threaded couplers — uses Slip-Loc quick-disconnect design
These samplers feature rigid 8-ft lengths of clear, 1-1/4”dia polycarbonate tubing which will not turn cloudy from sunlight or become brittle in cold weather like acrylic sludge samplers. The design offers a Slip-Loc quick-disconnect coupler and twin double o-ring seals to ensure a rugged, liquid-tight connection. Coretaker features a check valve for sample release; soft foam, floating lanyard handle; and pre-numbered yellow 1-ft markers.
43,100.00 - ₹79,500.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Length (ft)
Availability Pricing
Raven Environmental B-10104-8 Coretaker Polycarbonate Sludge Sampler, 8-ft 0546558 Mfr # B-10104-8
COMPARE
8
₹43,100.00INR / Each
Raven Environmental B-10104-16-REDESIGNED Coretaker Polycarbonate Sludge Sampler, 16-ft 0546560 Mfr # B-10104-16
COMPARE
16
₹48,400.00INR / Each
Raven Environmental B-10104-24 Coretaker Polycarbonate Sludge Sampler, 24-ft 0546562 Mfr # B-10104-24
COMPARE
24
₹79,500.00INR / Each