Radiation Alert Analog Radiation Detectors
Easy to use detector ensures safety
  • Precision sensitivity provides accurate readings
  • Easy setup and readability
All models are factory-calibrated to pulse generator and can be calibrated to ANSI standards by any certified lab. Anti-saturation circuit is good to at least 100 times the maximum reading in the highest range. No special probes are required. Ergonomic design decreases worker’s fatigue over long periods of use. Meters operate on one 9 V battery (sold separately).
171,000.00 - ₹177,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Function Range Description
Availability Pricing
Radiation Alert MONITOR 4EC Analog Radiation Detector, Energy-Compensated; 0-50 Mr/Hr 0898900 Mfr # MONITOR 4EC
COMPARE
Audible/visible counter 0 to 50 mR/hr; 0 to 50,000 cpm or 0 to 500 µSv/hr Analog Radiation Detector, Energy-Compensated; 0-50 Mr/Hr
₹177,000.00INR / Each
Radiation Alert MONITOR 4EC-USV Detector, Energy Compensated, 0 to 50 Mrad/Hr 0898930 Mfr # MONITOR 4EC-USV
COMPARE
Audible/visible counter 0 to 50 mR/hr; 0 to 50,000 cpm or 0 to 500 µSv/hr Detector, Energy Compensated, 0 to 50 Mrad/Hr
₹177,000.00INR / Each
Radiation Alert MONITOR 4, CPM Detector; 0 to 50 Mr/Hr, 0 to 50, 000 Cpm 0899050 Mfr # MONITOR 4 CPM
COMPARE
Audible/visible counter 0 to 50 mR/hr; 0 to 50,000 cpm or 0 to 500 µSv/hr Detector; 0 to 50 Mr/Hr, 0 to 50, 000 Cpm
₹171,000.00INR / Each