Qsonica Replacement Probes for the Touch-Screen Sonicator
Replacement Probe selection is determined by sample volume, sample type, and the desired intensity. As tip diameter increases, the intensity/amplitude decreases. Tips erode over time and replacement tips are available for order.
127,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Qsonica 4220 Sonicator Probe with Replaceable Tip; 1/2" Dia 0471266 Mfr # 4220
COMPARE
Sonicator Probe with Replaceable Tip; 1/2" Dia
₹127,000.00INR / Each
Qsonica 4207 Sonicator Probe with Replaceable Tip; 3/4" Dia 0471268 Mfr # 4207
COMPARE
Sonicator Probe with Replaceable Tip; 3/4" Dia
₹127,000.00INR / Each
Qsonica 4210 Sonicator Probe with Replaceable Tip; 1" Dia 0471270 Mfr # 4210
COMPARE
Sonicator Probe with Replaceable Tip; 1" Dia
₹127,000.00INR / Each