QRP Vinyl Cleanroom Gloves
Perfect for pharmaceuticals, electronics, and cleanroom food processing
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
Gloves feature a micro-textured surface for enhanced gripping. Latex-free, 100% PVC co-polymer is naturally antistatic with a resistivity of 1010 ohms. Beaded cuff for additional strength. Ambidextrous. Packed 100 gloves per polybag.
Compliance: ISO Class 5, USDA, and IEST-RP-CC-005.2.
8,800.00 - ₹9,100.00INR / Bag of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
QRP VHC12 SMALL Vinyl Cleanroom Gloves, small, 100/bag 8658020 Mfr # VHC12 SMALL
COMPARE
12 Small
₹8,800.00INR / Bag of 100
QRP VHC12 MEDIUM Vinyl Cleanroom Gloves, medium, 100/bag 8658021 Mfr # VHC12 MEDIUM
COMPARE
12 Medium
₹9,100.00INR / Bag of 100
QRP VHC12 LARGE Vinyl Cleanroom Gloves, large, 100/bag 8658022 Mfr # VHC12 LARGE
COMPARE
12 Large
₹9,100.00INR / Bag of 100