QRP Extreme-Temperature Gloves
Safely work with diffusion furnaces, liquid gas flasks, or hot/dry glassware
Knitted ESD gloves withstand dry contact temperatures with no charring or residue—will not contaminate annealing ovens or process equipment. Comfortable and flexible nylon lining keeps environment particulate free. Gloves are free of PCB, asbestos, and fiberglass.
Compliance: ISO Class 5; ESD per DODD-HDBCV-263 and MIL-B-81705.
74,000.00INR / Pair of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Size
Availability Pricing
QRP 50GL Extreme-Temperature Gloves, 14"L, Large 3368921 Mfr # 50GL
COMPARE
14 1400 Large
₹74,000.00INR / Pair of 1