QLA UltraCenter Precision Tapered Upper Spin Shafts
Significantly more accurate paddle and basket shafts
  • Tapered upper shaft, when fit to UltraCenter lower shafts, results in extremely concentric spin Interchangeable upper shafts add versatility and remove the need to readjust shaft height after changing Serialized—includes Certificate of Compliance (CoC)
Extremely concentric UltraCenter Precision tapered upper and lower shaft assembly provides more accurate spin without significant wobble. They were specifically designed to be used in concert with QLA AccuCenter™ brackets and UltraCenter™ vessels. The two-piece basket/paddle/cylinder shafts connect well above the mixing sample to prevent potential contamination. Constructed of highly bend-resistant 316 stainless steel and electropolished for smoothness and corrosion resistance.
These shafts are in compliance with the current USP and ASTM standards.
64,700.00 - ₹74,100.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Length (InchShort) Material Compatible With
Availability Pricing
QLA UltraCenter™ Precision Tapered Upper Spin Shaft for V-Series, 14.5", 316 SS; Serialized 4130113 Mfr # ASTMSHT-13
COMPARE
14 1/2 316 stainless steel Agilent/VanKel V-Series
₹64,700.00INR / Each
QLA UltraCenter™ Precision Tapered Upper Spin Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 15", 316 SS; Serialized 4130114 Mfr # ASTMSHT-15
COMPARE
15 316 stainless steel Agilent/VanKel/Varian
₹64,700.00INR / Each
QLA UltraCenter™ Precision Tapered Upper Spin Shaft for Hanson SR8-Plus, 16", 316 SS; Serialized 4130115 Mfr # ASTMSHT-16
COMPARE
16 316 stainless steel Hanson SR8-Plus
₹64,700.00INR / Each
QLA UltraCenter™ Precision Tapered Upper Spin Shaft for Distek 2100, 16.5", 316 SS; Serialized 4130116 Mfr # ASTMSHT-16.5
COMPARE
16 1/2 316 stainless steel Distek 2100
₹64,700.00INR / Each
QLA UltraCenter™ Precision Tapered Upper Spin Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 19", 316 SS; Serialized 4130117 Mfr # ASTMSHT-19
COMPARE
19 316 stainless steel Agilent/VanKel/Varian
₹69,600.00INR / Each
QLA UltraCenter™ Precision Tapered Upper Spin Shaft, 21", 316 SS; Serialized 4130118 Mfr # ASTMSHT-21
COMPARE
21 316 stainless steel Universal
₹71,600.00INR / Each
QLA UltraCenter™ Precision Tapered Upper Spin Shaft for Hanson Vision, 23", 316 SS; Serialized 4130119 Mfr # ASTMSHT-23
COMPARE
23 316 stainless steel Hanson Vision
₹73,100.00INR / Each
QLA UltraCenter™ Precision Tapered Upper Spin Shaft, 24", 316 SS; Serialized 4130120 Mfr # ASTMSHT-24
COMPARE
24 316 stainless steel Universal
₹74,100.00INR / Each
Showing 1- 8 of 8