QLA UltraCenter Precision Glass Vessels
Get better dissolution results with UltraCenter precision glass vessels
  • Glass offers good thermal conductivity and high resistance to scratching Serialized—include a Certificate of Conformance (CoC)
UltraCenter Precision glass vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius, resulting in more reproducible dissolution test results. They are compatible with most dissolution baths. Each vessel features a unique serial number.
121,000.00 - ₹181,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Bottle Color Special Features Description
Availability Pricing
QLA UltraCenter™ Precision Vessel, Amber Glass, 1000mL, with Plastic Rim; Serialized 4130103 Mfr # ASTM900-AUNP
COMPARE
Amber Plastic rim UltraCenter™ Precision Vessel, Amber Glass, 1000mL, with Plastic Rim; Serialized
₹164,000.00INR / Each
QLA UltraCenter™ Precision Vessel, Amber Glass, 1000mL, No Magnetic Collar for TruCenter; Serialized 4130104 Mfr # ASTM900-AVK
COMPARE
Amber Glass rim; no magnetic collar UltraCenter™ Precision Vessel, Amber Glass, 1000mL, No Magnetic Collar for TruCenter; Serialized
₹152,000.00INR / Each
QLA UltraCenter™ Precision Vessel, Clear Glass, 1000mL; Serialized 4130105 Mfr # ASTM900-UN
COMPARE
Clear Glass rim UltraCenter™ Precision Vessel, Clear Glass, 1000mL; Serialized
₹121,000.00INR / Each
QLA UltraCenter™ Precision Vessel, Amber Glass, 1000mL; Serialized 4130106 Mfr # ASTM900-UNAM
COMPARE
Amber Glass rim UltraCenter™ Precision Vessel, Amber Glass, 1000mL; Serialized
₹152,000.00INR / Each
QLA UltraCenter™ Precision Vessel, Clear Glass, 1000mL, with Plastic Rim; Serialized 4130107 Mfr # ASTM900-UNP
COMPARE
Clear Plastic rim UltraCenter™ Precision Vessel, Clear Glass, 1000mL, with Plastic Rim; Serialized
₹147,000.00INR / Each
QLA UltraCenter™ Precision Vessel, Clear Glass, 1000mL, for TruCenter, No Magnetic Collar; Serialized 4130108 Mfr # ASTM900-VK
COMPARE
Clear Glass rim; no magnetic collar UltraCenter™ Precision Vessel, Clear Glass, 1000mL, for TruCenter, No Magnetic Collar; Serialized
₹121,000.00INR / Each
QLA UltraCenter™ Precision PEAK Vessel, Amber Glass, 1000mL, with Plastic Rim; Serialized 4130111 Mfr # ASTM9PK-AUNP
COMPARE
Clear Plastic rim; PEAK bottom UltraCenter™ Precision PEAK Vessel, Amber Glass, 1000mL, with Plastic Rim; Serialized
₹181,000.00INR / Each
QLA UltraCenter™ Precision PEAK Vessel, Clear Glass, 1000mL, with Plastic Rim; Serialized 4130112 Mfr # ASTM9PK-UNP
COMPARE
Amber Plastic rim; PEAK bottom UltraCenter™ Precision PEAK Vessel, Clear Glass, 1000mL, with Plastic Rim; Serialized
₹170,000.00INR / Each
Showing 1- 8 of 8