QLA Porous Cannula Filters for Sotax Baths
Achieve reproducibility and consistency from test to test with QLA dissolution filters
  • Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds and provide a good flow rate.
29,800.00 - ₹408,000.00INR / Pkg of 1000
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Internal Diameter (mm) Pore Size (micron) Description
Availability Pricing
QLA Porous Filters, 10 µm, UHMW PE, 1/8" ID, Sotax compatible; 1000/PK 4130910 Mfr # FIL010-ST
COMPARE
3.2 10 Porous Filters, 10 µm, UHMW PE, 1/8" ID, Sotax compatible; 1000/PK
₹254,000.00INR / Pkg of 1000
QLA Porous Filters, 10 µm, UHMW PE, 1/8" ID, Sotax compatible; 100/PK 4130911 Mfr # FIL010-ST-A
COMPARE
3.2 10 Porous Filters, 10 µm, UHMW PE, 1/8" ID, Sotax compatible; 100/PK
₹29,800.00INR / Pkg of 100
QLA Porous Filters, 20 µm, UHMW PE, 1/8" ID, Sotax compatible; 1000/PK 4130924 Mfr # FIL020-ST
COMPARE
3.2 20 Porous Filters, 20 µm, UHMW PE, 1/8" ID, Sotax compatible; 1000/PK
₹307,000.00INR / Pkg of 1000
QLA Porous Filters, 20 µm, UHMW PE, 1/8" ID, Sotax compatible; 100/PK 4130925 Mfr # FIL020-ST-a
COMPARE
3.2 20 Porous Filters, 20 µm, UHMW PE, 1/8" ID, Sotax compatible; 100/PK
₹35,800.00INR / Pkg of 100
QLA Porous Filters, 35 µm, UHMW PE, 1/8" ID, Sotax compatible; 1000/PK 4130939 Mfr # FIL035-ST
COMPARE
3.2 35 Porous Filters, 35 µm, UHMW PE, 1/8" ID, Sotax compatible; 1000/PK
₹329,000.00INR / Pkg of 1000
QLA Porous Filters, 35 µm, UHMW PE, 1/8" ID, Sotax compatible; 100/PK 4130940 Mfr # FIL035-ST-A
COMPARE
3.2 35 Porous Filters, 35 µm, UHMW PE, 1/8" ID, Sotax compatible; 100/PK
₹38,500.00INR / Pkg of 100
QLA Porous Filters, 70 µm, UHMW PE, 1/8" ID, Sotax compatible; 1000/PK 4130967 Mfr # FIL070-ST
COMPARE
3.2 70 Porous Filters, 70 µm, UHMW PE, 1/8" ID, Sotax compatible; 1000/PK
₹365,000.00INR / Pkg of 1000
QLA Porous Filters, 70 µm, UHMW PE, 1/8" ID, Sotax compatible; 100/PK 4130968 Mfr # FIL070-ST-A
COMPARE
3.2 70 Porous Filters, 70 µm, UHMW PE, 1/8" ID, Sotax compatible; 100/PK
₹38,500.00INR / Pkg of 100
QLA Porous Filters, 10 µm, UHMW PE, 1/16" ID, Sotax compatible; 1000/PK 4130977 Mfr # FIL10S-ST
COMPARE
1.6 10 Porous Filters, 10 µm, UHMW PE, 1/16" ID, Sotax compatible; 1000/PK
₹408,000.00INR / Pkg of 1000
Showing 1- 9 of 9