QLA Porous Cannula Filters for Erweka Baths
Achieve reproducibility and consistency from test to test with QLA dissolution filters
  • Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds and provide a good flow rate.
29,800.00 - ₹351,000.00INR / Pkg of 1000
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (micron) Description
Availability Pricing
QLA Porous Filters, 1 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK 4130878 Mfr # FIL001-EW
COMPARE
1 Porous Filters, 1 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK
₹330,000.00INR / Pkg of 1000
QLA Porous Filters, 1 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK 4130879 Mfr # FIL001-EW-A
COMPARE
1 Porous Filters, 1 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK
₹45,700.00INR / Pkg of 100
QLA Porous Filters, 4 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK 4130888 Mfr # FIL004-EW
COMPARE
4 Porous Filters, 4 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK
₹330,000.00INR / Pkg of 1000
QLA Porous Filters, 4 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK 4130889 Mfr # FIL004-EW-A
COMPARE
4 Porous Filters, 4 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK
₹45,700.00INR / Pkg of 100
QLA Porous Filters, 10 µm,  UHMW PE, 1/8" ID, Erweka compatible; 1000/PK 4130904 Mfr # FIL010-EW
COMPARE
10 Porous Filters, 10 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK
₹254,000.00INR / Pkg of 1000
QLA Porous Filters, 10 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK 4130905 Mfr # FIL010-EW-A
COMPARE
10 Porous Filters, 10 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK
₹29,800.00INR / Pkg of 100
QLA Porous Filters, 20 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK 4130918 Mfr # FIL020-EW
COMPARE
20 Porous Filters, 20 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK
₹307,000.00INR / Pkg of 1000
QLA Porous Filters, 20 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK 4130919 Mfr # FIL020-EW-A
COMPARE
20 Porous Filters, 20 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK
₹35,800.00INR / Pkg of 100
QLA Porous Filters, 35 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK 4130933 Mfr # FIL035-EW
COMPARE
35 Porous Filters, 35 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK
₹258,000.00INR / Pkg of 1000
QLA Porous Filters, 35 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK 4130934 Mfr # FIL035-EW-a
COMPARE
35 Porous Filters, 35 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK
₹31,200.00INR / Pkg of 100
QLA Porous Filters, 40 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK 4130945 Mfr # FIL040-EW
COMPARE
40 Porous Filters, 40 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK
₹263,000.00INR / Pkg of 1000
QLA Porous Filters, 40 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK 4130946 Mfr # FIL040-EW-A
COMPARE
40 Porous Filters, 40 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK
₹38,500.00INR / Pkg of 100
QLA Porous Filters, 70 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK 4130961 Mfr # FIL070-EW
COMPARE
70 Porous Filters, 70 µm, UHMW PE, 1/8" ID, Erweka compatible; 1000/PK
₹351,000.00INR / Pkg of 1000
QLA Porous Filters, 70 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK 4130962 Mfr # FIL070-EW-A
COMPARE
70 Porous Filters, 70 µm, UHMW PE, 1/8" ID, Erweka compatible; 100/PK
₹58,100.00INR / Pkg of 100
Showing 1- 14 of 14