QLA Porous Cannula Filters for Distek Baths
Achieve reproducibility and consistency from test to test with QLA dissolution filters
  • Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds and provide a good flow rate.
48,300.00 - ₹347,000.00INR / Pkg of 1000
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (micron) Description
Availability Pricing
QLA Porous Filters, 10 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK 4130973 Mfr # FIL10S-DK
COMPARE
10 Porous Filters, 10 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK
₹347,000.00INR / Pkg of 1000
QLA Porous Filters, 10 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK 4130974 Mfr # FIL10S-DK-A
COMPARE
10 Porous Filters, 10 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK
₹48,300.00INR / Pkg of 100
QLA Porous Filters, 20 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK 4130978 Mfr # FIL20S-DK
COMPARE
20 Porous Filters, 20 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK
₹347,000.00INR / Pkg of 1000
QLA Porous Filters, 20 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK 4130979 Mfr # FIL20S-DK-A
COMPARE
20 Porous Filters, 20 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK
₹48,300.00INR / Pkg of 100
QLA Porous Filters, 35 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK 4130982 Mfr # FIL35S-DK
COMPARE
35 Porous Filters, 35 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK
₹347,000.00INR / Pkg of 1000
QLA Porous Filters, 35 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK 4130983 Mfr # FIL35S-DK-A
COMPARE
35 Porous Filters, 35 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK
₹48,300.00INR / Pkg of 100
QLA Porous Filters, 45 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK 4130986 Mfr # FIL45S-DK
COMPARE
45 Porous Filters, 45 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK
₹347,000.00INR / Pkg of 1000
QLA Porous Filters, 45 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK 4130987 Mfr # FIL45S-DK-A
COMPARE
45 Porous Filters, 45 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK
₹48,300.00INR / Pkg of 100
QLA Porous Filters, 70 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK 4130990 Mfr # FIL70S-DK
COMPARE
70 Porous Filters, 70 µm, UHMW PE, 1/16" ID, Distek compatible; 1000/PK
₹347,000.00INR / Pkg of 1000
QLA Porous Filters, 70 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK 4130991 Mfr # FIL70S-DK-a
COMPARE
70 Porous Filters, 70 µm, UHMW PE, 1/16" ID, Distek compatible; 100/PK
₹55,800.00INR / Pkg of 100
Showing 1- 10 of 10