QLA Porous Cannula Filters for Caleva Baths
Achieve reproducibility and consistency from test to test with QLA dissolution filters
  • Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds and provide a good flow rate.
29,800.00 - ₹330,000.00INR / Pkg of 1000
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (micron) Description
Availability Pricing
QLA Porous Filters, 1 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK 4130876 Mfr # FIL001-CA
COMPARE
1 Porous Filters, 1 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK
₹330,000.00INR / Pkg of 1000
QLA Porous Filters, 1 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK 4130877 Mfr # FIL001-CA-A
COMPARE
1 Porous Filters, 1 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK
₹45,700.00INR / Pkg of 100
QLA Porous Filters, 4 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK 4130886 Mfr # FIL004-CA
COMPARE
4 Porous Filters, 4 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK
₹330,000.00INR / Pkg of 1000
QLA Porous Filters, 4 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK 4130887 Mfr # FIL004-CA-A
COMPARE
4 Porous Filters, 4 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK
₹45,700.00INR / Pkg of 100
QLA Porous Filters, 10 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK 4130900 Mfr # FIL010-CA
COMPARE
10 Porous Filters, 10 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK
₹254,000.00INR / Pkg of 1000
QLA Porous Filters, 10 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK 4130901 Mfr # FIL010-CA-A
COMPARE
10 Porous Filters, 10 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK
₹29,800.00INR / Pkg of 100
QLA Porous Filters, 35 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK 4130929 Mfr # FIL035-CA
COMPARE
35 Porous Filters, 35 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK
₹258,000.00INR / Pkg of 1000
QLA Porous Filters, 35 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK 4130930 Mfr # FIL035-CA-A
COMPARE
35 Porous Filters, 35 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK
₹31,200.00INR / Pkg of 100
QLA Porous Filters, 40 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK 4130943 Mfr # FIL040-CA
COMPARE
40 Porous Filters, 40 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK
₹263,000.00INR / Pkg of 1000
QLA Porous Filters, 40 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK 4130944 Mfr # FIL040-CA-a
COMPARE
40 Porous Filters, 40 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK
₹38,500.00INR / Pkg of 100
QLA Porous Filters, 70 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK 4130957 Mfr # FIL070-CA
COMPARE
70 Porous Filters, 70 µm, UHMW PE, 1/8" ID, Caleva compatible; 1000/PK
₹263,000.00INR / Pkg of 1000
QLA Porous Filters, 70 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK 4130958 Mfr # FIL070-CA-a
COMPARE
70 Porous Filters, 70 µm, UHMW PE, 1/8" ID, Caleva compatible; 100/PK
₹38,500.00INR / Pkg of 100
Showing 1- 12 of 12