QLA Full-Flow, Direct Fit Cannula Filters Compatible with Agilent and Legacy VanKel Baths
Achieve reproducibility and consistency from test to test with QLA dissolution filters
  • No Tygon® tubing needed Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. These direct fit filters fit directly onto the end of the standard cannula and do not require any tubing. Filters are completely inert to most compounds and provide a good flow rate.
27,800.00 - ₹263,000.00INR / Pkg of 1000
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (micron) Description
Availability Pricing
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 10 µm, UHMW PE; 1000/PK 4130912 Mfr # FIL010-VK
COMPARE
10 Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 10 µm, UHMW PE; 1000/PK
₹263,000.00INR / Pkg of 1000
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 10 µm, UHMW PE; 100/PK 4130913 Mfr # FIL010-VK-A
COMPARE
10 Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 10 µm, UHMW PE; 100/PK
₹30,800.00INR / Pkg of 100
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 35 µm, UHMW PE; 1000/PK 4130941 Mfr # FIL035-VK
COMPARE
35 Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 35 µm, UHMW PE; 1000/PK
₹263,000.00INR / Pkg of 1000
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 35 µm, UHMW PE; 100/PK 4130942 Mfr # FIL035-VK-A
COMPARE
35 Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 35 µm, UHMW PE; 100/PK
₹31,200.00INR / Pkg of 100
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 45 µm, UHMW PE; 1000/PK 4130953 Mfr # FIL045-VK
COMPARE
45 Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 45 µm, UHMW PE; 1000/PK
₹263,000.00INR / Pkg of 1000
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 45 µm, UHMW PE; 100/PK 4130954 Mfr # FIL045-VK-a
COMPARE
45 Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 45 µm, UHMW PE; 100/PK
₹37,500.00INR / Pkg of 100
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 70 µm, UHMW PE; 1000/PK 4130969 Mfr # FIL070-VK
COMPARE
70 Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 70 µm, UHMW PE; 1000/PK
₹239,000.00INR / Pkg of 1000
QLA Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 70 µm, UHMW PE; 100/PK 4130970 Mfr # FIL070-VK-A
COMPARE
70 Full-Flow Filters Compatible with Agilent and VanKel Dissolution Apparatus, Direct Fit, 70 µm, UHMW PE; 100/PK
₹27,800.00INR / Pkg of 100
Showing 1- 8 of 8