QLA Erweka-Compatible Baskets
QLA Baskets for dissolution testing with your Erweka apparatus
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Erweka test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
36,800.00 - ₹175,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Mesh Size Description
Availability Pricing
QLA Clip Style Basket for Erweka, 10 Mesh, 316 SS; Serialized 4130131 Mfr # BSK010-EW
COMPARE
316 stainless steel 10 Clip Style Basket for Erweka, 10 Mesh, 316 SS; Serialized
₹45,000.00INR / Each
QLA Clip Style Basket for Erweka, 20 Mesh, 316 SS; Serialized 4130139 Mfr # BSK020-EW
COMPARE
316 stainless steel 20 Clip Style Basket for Erweka, 20 Mesh, 316 SS; Serialized
₹51,200.00INR / Each
QLA Clip Style Basket for Erweka, 40 Mesh USP, 316 SS; Serialized 4130149 Mfr # BSK040-EW
COMPARE
316 stainless steel 40 Clip Style Basket for Erweka, 40 Mesh USP, 316 SS; Serialized
₹36,800.00INR / Each
QLA Clip Style Basket for Erweka, 150 Mesh, 316 SS; Serialized 4130167 Mfr # BSK150-EW
COMPARE
316 stainless steel 150 Clip Style Basket for Erweka, 150 Mesh, 316 SS; Serialized
₹150,000.00INR / Each
QLA Suppository Basket for Erweka; Plastic 4130195 Mfr # BSKSUP-EW
COMPARE
Plastic Suppository Basket for Erweka; Plastic
₹55,500.00INR / Each
QLA Clip Style Basket for Erweka, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130842 Mfr # CUSBSK-20/100EW
COMPARE
316 stainless steel 100 (plus 20 mesh stability lining) Clip Style Basket for Erweka, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
QLA Clip Style Basket for Erweka, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130849 Mfr # CUSBSK-20/150EW
COMPARE
316 stainless steel 150 (plus 20 mesh stability lining) Clip Style Basket for Erweka, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
Showing 1- 7 of 7