QLA Distek-Compatible Dissolution Vessels
QLA vessels provide more precise and consistent dissolution results
  • 1000 mL capacity
Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle. Both Standard and PEAK vessels come with an Acculign ring to help center the vessel. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.
49,600.00 - ₹84,100.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Bottle Color Material Special Features
Availability Pricing
QLA Standard Vessel with Acculign Ring for Distek, 1000 mL, Amber Glass; Serialized 4130643 Mfr # GLA9CR-AD
COMPARE
Amber Glass Standard with Acculign ring
₹84,100.00INR / Each
QLA Standard Vessel with Acculign Ring for Distek, 1000 mL, Clear Glass; Serialized 4130645 Mfr # GLA9CR-DK
COMPARE
Clear Glass Standard with Acculign ring
₹54,000.00INR / Each
QLA PEAK Vessel with Acculign Ring for Distek, 1000 mL, Clear Glass; Serialized 4130653 Mfr # GLA9PK-DK
COMPARE
Clear Glass PEAK-style with Acculign ring
₹77,000.00INR / Each
QLA Footed Vessel for Distek, 1000 mL; Amber Plastic 4130657 Mfr # PLA900-AD
COMPARE
Amber Plastic Footed
₹54,700.00INR / Each
QLA Footed Vessel for Distek, 1000 mL; Clear Plastic 4130661 Mfr # PLA900-DK
COMPARE
Clear Plastic Footed
₹49,600.00INR / Each