Pyrex Brand 9990 Watch Glasses
Ribbed, beaker cover
Use these PYREX watch glasses to perform evaporations and as a cover for your beakers. Ribs are molded into the glass, permitting reflux of digestate and escape of excess vapor. Meets EPA requirement for methods 3005, 3010, and 3020.
21,900.00 - ₹37,400.00INR / Case of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fits Bottle / Beaker Size (mL) Material Outside Diameter (OD) (cm)
Availability Pricing
Pyrex 9990-75 Brand 9990 ribbed watch glass; 75 mm, case of 12 WW-34508-21 Mfr # 9990-75
COMPARE
250 Glass 7.5
₹21,900.00INR / Case of 12
Pyrex 9990-100 Brand 9990 ribbed watch glass; 600 mm, case of 12 WW-34508-22 Mfr # 9990-100
COMPARE
600 Glass 10
₹26,800.00INR / Case of 12
Pyrex 9990-125 Brand 9990 ribbed watch glass; 1000 mm, case of 12 WW-34508-23 Mfr # 9990-125
COMPARE
1000 Glass 12.5
₹37,400.00INR / Case of 12