Pyrex Brand 3026 Cylinders
  • Double metric scale
  • Class A
These graduated cylinders are calibrated “to deliver.” Double metric scale features white or blue enamel graduations. Blue enamel markings are ideal for milky white liquids. Shock absorbing polyethylene bumper guards are supplied. Reference: ASTM E-1272 and ASTM E-542.
19,600.00 - ₹102,000.00INR / Case of 18
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex 3026-50 Brand 3026 Cylinder, Class A, To Deliver, 50 ml, 18/cs WW-34592-22 Mfr # 3026-50
COMPARE
50 1 Glass
₹102,000.00INR / Case of 18
Pyrex 3026-1L Brand 3026 Cylinder, Class A, To Deliver, 1000 mL, 1/cs WW-34592-26 Mfr # 3026-1L
COMPARE
1000 10 Glass
₹19,600.00INR / Case of 1
Pyrex 3026-2L Brand 3026 Cylinder, Class A, To Deliver, 2000 mL, 1/cs WW-34592-27 Mfr # 3026-2L
COMPARE
2000 20 Glass
₹31,900.00INR / Case of 1