Pulse-Free Regenerative Blowers
Sound-absorbing design minimizes air noise
  • Runs dry
  • TEFC motor-mounted
  • Special sealing minimizes air leakage
  • All-aluminum construction reduces weight and results in uniform thermal expansions
  • Most models have sound emissions lower than OSHA requirements
  • Three-year parts and workmanship warranty
Regenerative blowers are ideal for pond or plate bath aeration, wastewater treatment, vacuum lifting, trim removal, pneumatic conveying, and dust/smoke removal. Blowers provide oilless, pulse-free air at a very low decibel rating of 58 to 80 dB(A). Unique internal design channels air into sound-absorbing foam to minimize air noise—most models have sound emissions lower than OSHA requirements. Blowers can be mounted in any plane.
175,000.00 - ₹960,000.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Power (Hz) Power (VAC)
Availability Pricing
FPZ SCL-06B Regenerative Blowers, 39 cfm (1104 L/min), 115/230 VAC 7960710 Mfr # SCL 06B-.33-115/230 NP
COMPARE
39 60 115 / 230
₹175,000.00INR / Each
FPZ SCL-K03 Regenerative Blowers, 49 cfm (1387 L/min), 115/230 VAC, 1 Ph 7960720 Mfr # SCL K03-MS-.75-115/230 NP
COMPARE
52 60 115 / 230
₹339,000.00INR / Each
FPZ SCL-K03 Regenerative Blowers, 49 cfm (1387 L/min), 230/460 VAC, 3 Ph 7960730 Mfr # SCL K03-MS-.75-3 NP
COMPARE
52 60/50 230 / 460
₹221,000.00INR / Each
FPZ SCL 40D Regenerative Blowers, 98 cfm (2775 L/min), 208-230/460 VAC, 4 Hp 7960736 Mfr # SCL R40-MD-4-3
COMPARE
98 60/50 230 / 460
₹634,000.00INR / Each
FPZ SCL-K04 Regenerative Blowers, 98 cfm (2690 L/min), 230 VAC 7960740 Mfr # SCL K04-MS-2-115/230 NP
COMPARE
98 60 115 / 230
₹365,000.00INR / Each
FPZ SCL-K04 Regenerative Blowers, 98 cfm (2690 L/min), 230/460 VAC 7960750 Mfr # SCL K04-MS-2-3 NP
COMPARE
98 60/50 230 / 460
₹294,000.00INR / Each
FPZ SCL K04 Regenerative Blowers, 98 cfm (2775 L/min), 208-230/460 VAC, 3 Hp 7960753 Mfr # SCL K04-MS-3-3 NP
COMPARE
98 60/50 230 / 460
₹291,000.00INR / Each
FPZ SCL-K05 Regenerative Blowers, 156 cfm (4417 L/min), 230/460 VAC 7960760 Mfr # SCL K05-MS-3-3 NP
COMPARE
156 60/50 230 / 460
₹362,000.00INR / Each
FPZ SCL K05 Regenerative Blowers, 156 cfm (4417 L/min), 208-230/460 VAC, 4 Hp 7960763 Mfr # SCL K05-MS-4-3 NP
COMPARE
156 60/50 230 / 460
₹426,000.00INR / Each
FPZ SCL-K06 Regenerative Blowers, 213 cfm (6031 L/min), 230/460 VAC 7960770 Mfr # SCL K06-MS-5.5-3 NP
COMPARE
216 60/50 230 / 460
₹461,000.00INR / Each
FPZ SCL K07 Regenerative Blowers, 294 cfm (8325 L/min), 208-230/460 VAC, 10 Hp 7960773 Mfr # SCL K07-MS-10-3
COMPARE
294 60/50 230 / 460
₹960,000.00INR / Each
FPZ SCL E08-MS Regenerative Blowers, 381 cfm (10789 L/min), 230/460 VAC 7960780 Mfr # SCL-E08-MS
COMPARE
381 60/50 230 / 460
Call For Price
FPZ SCL E10-MS Regenerative Blowers, 584 cfm (16,536 L/min), 208-230/460 VAC, 20 Hp 7960783 Mfr # SCL E10-MS
COMPARE
584 60/50 230 / 460
Call For Price
Showing 1- 13 of 13