Powerblanket® Lite Drum and Pail Heaters
Conserve energy and save money while maintaining optimal temperature
  • Maintenance free heating solution
  • Prevent temperature runaway with built-in preset
  • Install in minutes by simply wrapping the heater around the vessel
The Lite series drum and bucket heaters feature great insulation and heating uniformity for your less rugged applications. Avoid excess power usage and conserve energy while still quickly heating temperature sensitive materials.
Designed with a thin outer layer to keep heat contained, these heaters direct energy inward to provide quick and efficient heating. Achieve even heat distribution with the full wrap around design, eliminating hot or cold spots and scorching or burning.
All models feature a preset thermostat set for maximum product temperatures. An adjustable, thermostatic controller is sold separately as part number 42025-15.
47,500.00 - ₹97,700.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Size Max Temperature (° F) Description
Availability Pricing
Powerblanket PBL05 Lite Bucket Heater, 5 Gallon; 120 V 4202529 Mfr # PBL05
COMPARE
5 gallons 145 Powerblanket Lite Bucket Heater, 5 Gallon; 120 V
₹52,200.00INR / Each
Powerblanket PBL15 Lite Bucket Heater, 15 Gallon; 120 V 4202530 Mfr # PBL15
COMPARE
15 gallons 145 Powerblanket Lite Bucket Heater, 15 Gallon; 120 V
₹62,700.00INR / Each
Powerblanket PBL30 Lite Drum Heater, 30 Gallon; 120 V 4202531 Mfr # PBL30
COMPARE
30 gallons 145 Powerblanket Lite Drum Heater, 30 Gallon; 120 V
₹68,500.00INR / Each
Powerblanket PBL55 Lite Drum Heater, 55 Gallon; 120 V 4202532 Mfr # PBL55
COMPARE
55 gallons 145 Powerblanket Lite Drum Heater, 55 Gallon; 120 V
₹97,700.00INR / Each
Powerblanket PBL1G Lite Pail Heater, 1 Gallon; 120 V 4202533 Mfr # PBL1G
COMPARE
1 gallon 84 Powerblanket Lite Pail Heater, 1 Gallon; 120 V
₹47,500.00INR / Each
Powerblanket PBL2G Lite Pail Heater, 2 Gallon; 120 V 4202534 Mfr # PBL2G
COMPARE
2 gallons 84 Powerblanket Lite Pail Heater, 2 Gallon; 120 V
₹48,600.00INR / Each