Powerblanket® Gas Cylinder Heaters

Increase flow rate performance and efficiency
  • Quickly heats cylinder to a preset 90°F (32°C)
  • Eliminates cylinder refills in cold weather
  • Easy to install and remove
  • Certified to UL/CSA/CE standards
The gas cylinder heaters allow you to overcome the effects of cold weather, preventing fluids from freezing while maintaining optimal temperatures. The unique insulated full-wrap design delivers even heat distribution, helping to optimize the container pressure and increase the cylinder efficiency.
68,500.00 - ₹234,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Size Length (in) Power (watts)
Availability Pricing
Powerblanket PBL20 Lite Gas Cylinder Heater, 20 lbs 4202544 Mfr # PBL20
COMPARE
20 to 40 lbs 46 120
₹68,500.00INR / Each
Powerblanket PBL100 Lite Gas Cylinder Heater, 100 lbs 4202545 Mfr # PBL100
COMPARE
100 lbs 55 280
₹104,000.00INR / Each
Powerblanket PBL420 Lite Gas Cylinder Heater, 420 lbs 4202546 Mfr # PBL420
COMPARE
420 lbs 100 1/2 400
₹134,000.00INR / Each
Powerblanket GCW40 Gas Cylinder Heater, 40 lbs 4202549 Mfr # GCW40
COMPARE
40 lbs 45 280
₹198,000.00INR / Each
Powerblanket GCW100 Gas Cylinder Heater, 100 lbs 4202550 Mfr # GCW100
COMPARE
100 lbs 52 550
₹234,000.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat