Powerblanket® Drum and Pail Heating Blankets
Full-wrap design ensures even heat distribution no matter the size of the drum
  • Full insulation protects against freezing temperatures
  • Suitable for use on poly and steel drums
  • Delivers evenly distributed heat, eliminating hot or cold spots
112,000.00 - ₹300,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Size Max Temperature (° F) Power (watts)
Availability Pricing
Powerblanket BH05PRO Pro Pail Heater, 5 Gallon; 120 VAC 4202500 Mfr # BH05PRO
COMPARE
5 gal. 145 160
₹176,000.00INR / Each
Powerblanket BH05RR Rapid Ramp Pail Heater, 5 Gallon; 120 VAC 4202501 Mfr # BH05RR
COMPARE
5 gal. 100 120
₹112,000.00INR / Each
Powerblanket BH15PRO Pro Drum Heater, 15 Gallon; 120 VAC 4202502 Mfr # BH15PRO
COMPARE
15 gal. 145 400
₹234,000.00INR / Each
Powerblanket BH30PRO Pro Drum Heater, 30 Gallon; 120 VAC 4202504 Mfr # BH30PRO
COMPARE
30 gal. 145 720
₹268,000.00INR / Each
Powerblanket BH30RR Rapid Ramp Drum Heater, 30 Gallon; 120 VAC 4202505 Mfr # BH30RR
COMPARE
30 gal. 100 560
₹167,000.00INR / Each
Powerblanket BH55PRO Pro Drum Heater, 55 Gallon; 120 VAC 4202506 Mfr # BH55PRO
COMPARE
55 gal. 145 800
₹300,000.00INR / Each
Powerblanket BH55RR100 Rapid Ramp Drum Heater, 55 Gallon, 100 °F; 120 VAC 4202507 Mfr # BH55RR100
COMPARE
55 gal. 100 800
₹203,000.00INR / Each
Powerblanket BH55RR120 Rapid Ramp Drum Heater, 55 Gallon, 120 °F; 120 VAC 4202508 Mfr # BH55RR120
COMPARE
55 gal. 120 800
₹203,000.00INR / Each
Powerblanket BH55RR80 Rapid Ramp Drum Heater, 55 Gallon, 80 °F; 120 VAC 4202509 Mfr # BH55RR80
COMPARE
55 gal. 80 800
₹203,000.00INR / Each
Showing 1- 9 of 9