Polyethylene-Coated Polypropylene Lab Coats
Excellent lightweight splash protection
These polyethylene (PE) coated polypropylene (PP) lab coats are a low-cost disposable alternative to Tyvek® coats. The durable, lightweight, and breathable fabric is ideal for general-purpose laboratory applications. The PE-coated spun bond PP-provides excellent splash protection from water-based liquids.
21,200.00 - ₹22,400.00INR / Case of 30
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Size
Availability Pricing
Total Source APP0242-W Polyethylene-Coated Polypropylene Lab Coats, Medium, case of 30 8622712 Mfr # APP0242-W-M
COMPARE
White 41.5 Medium
₹21,200.00INR / Case of 30
Total Source APP0242-W Polyethylene-Coated Polypropylene Lab Coats, Large, case of 30 8622714 Mfr # APP0242-W-L
COMPARE
White 41.5 Large
₹22,400.00INR / Case of 30