Polyethylene Cleanroom Notebooks

Polyethylene-binding technology reduces paper fiber and allows recycling
Nonlatex cleanroom paper is impregnated with polyethylene and doesn't use organic fillers — paper is 100% recyclable. Paper is produced in an ISO Class 5 (formerly Class 100) environment and cleaned and double bagged in an ISO Class 4 (formerly Class 10) environment. Features excellent printability and heat toner adhesion, with high opacity and durability.
44,300.00 - ₹53,500.00INR / Case of 20

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Width (in) Description
Availability Pricing
Purus 08NBP-8.5x11 Polyethylene Cleanroom Notebook, 8-1/2" x 11", 100 page, 28 lb., 10/cs 3367862 Mfr # PNB CI 8.5X11 2210-L
COMPARE
8.5 11 Polyethylene Cleanroom Notebook, 8-1/2" x 11", 100 page, 28 lb., 10/cs
₹44,300.00INR / Case of 10
Purus 08NBP-5.5x8 Polyethylene Cleanroom Notebook, 5-1/2" x 8-1/2", 100 page, 28 lb., 20/cs 3367863 Mfr # PNB CI 5.5X8.5 2220-L
COMPARE
5.5 8.5 Polyethylene Cleanroom Notebook, 5-1/2" x 8-1/2", 100 page, 28 lb., 20/cs
₹53,500.00INR / Case of 20
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat