Pocket Durometers
Unbreakable, lightweight, and easy to carry around the plant for on-site testing
  • Scale is divided into 5-point increments from 0 to 100
The sliding vernier scale is easy to read and has no cams, gears, or levers to wear out. Scale holds maximum reading making it ideal for testing hard-to-reach areas. Measure 1/2"W x 4-1/2"H.
57,300.00 - ₹82,500.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Calibration Included Type
Availability Pricing
Rex Gauge 1500 TYPE A Pocket Hardness Tester/Durometer; Type A 5979200 Mfr # 1500 TYPE A
COMPARE
No A
₹57,300.00INR / Each
Rex Gauge 1500 TYPE D Pocket Hardness Tester/Durometer, Type D 5979202 Mfr # 1500 TYPE D
COMPARE
No D
₹63,800.00INR / Each
Rex Gauge 1500 TYPE A Pocket Durometer with NIST Calibration Certificate, Type A 5979228 Mfr # 1500 TYPE A W/NIST CERT
COMPARE
Yes A
₹82,500.00INR / Each
Rex Gauge 1500 TYPE D Pocket Durometer with NIST Calibration Certificate, Type D 5979230 Mfr # 1500 TYPE D W/NIST CERT
COMPARE
Yes D
₹76,500.00INR / Each