Plast-O-Matic 2-Way Sanitary Ball Valves, Polypropylene
Ensure easy clean, leak-free installation with sanitary connections
  • Mirror-polished, concentric ball provides leakproof operation
  • PTFE bearing and seats offer smooth turning
  • Dual shaft EPDM seals eliminate leaks
  • Top and bottom trunnion design securely holds ball in place
  • Ultra smooth flow path minimizes turbulence and pressure loss
  • Natural polypropylene does not contain any plasticizers or fillers and offers good chemical resistance and impact strength
Easily control fluid flow with the 2-way ball valves. With a simple turn, these valves allow the through-hole configuration to be blocked, open, or partially open to regulate the flow. Ball valves feature a fully concentric, mirror-polished ball to create a tight seal, eliminating any leakage when closed. PTFE bearing on shaft and seats ensure smooth turning for ease of use and longer cycle life. The ultra smooth flow path ensures minimal turbulence and pressure loss, ensuring excellent flow rates. A heavy-duty, large diameter shaft eliminates flexing and breakage while top and bottom trunnion design minimizes lateral ball stress for excellent durability. Easily connect sanitary ball valves to existing systems with Tri-Clamp or Tri-Clover fittings.
68,700.00 - ₹131,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Flow Coefficient (Cv) Inlet Port Outlet Port
Availability Pricing
Plast-O-Matic 2-Way Sanitary Ball Valve, PP, 1/2" Mini 4852433 Mfr # MBV050EPSC-PP
COMPARE
10 1/2" Mini 1/2" Mini
₹68,700.00INR / Each
Plast-O-Matic 2-Way Sanitary Ball Valve, PP, 3/4" Mini 4852434 Mfr # MBV075EPSC-PP
COMPARE
20 3/4" Mini 3/4" Mini
₹75,800.00INR / Each
Plast-O-Matic 2-Way Sanitary Ball Valve, PP, 1" Maxi 4852435 Mfr # MBV100EPSCM-PP
COMPARE
40 1" Maxi 1" Maxi
₹87,100.00INR / Each
Plast-O-Matic 2-Way Sanitary Ball Valve, PP, 1" Ladish 4852436 Mfr # MBV100EPSC-PP
COMPARE
40 1" Ladish 1" Ladish
₹90,200.00INR / Each
Plast-O-Matic 2-Way Sanitary Ball Valve, PP, 1.5" 4852437 Mfr # MBV150EPSC-PP
COMPARE
100 1.5" 1.5"
₹110,000.00INR / Each
Plast-O-Matic 2-Way Sanitary Ball Valve, PP, 2" 4852438 Mfr # MBV200EPSC-PP
COMPARE
120 2" 2"
₹131,000.00INR / Each