PharmaFluor® Panel-Mount Compression Fittings, Straight Union, PFA
High-purity compression fittings designed for critical fluid and gas applications
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
17,900.00 - ₹23,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Flow Pattern
Availability Pricing
PharmaFluor Panel-Mount Compression Fitting, Straight Union, PFA, 1/4" OD 5014435 Mfr # G1077013
COMPARE
1/4 1/4 Straight
₹17,900.00INR / Each
PharmaFluor Panel-Mount Compression Fitting, Straight Union, PFA, 1/8" OD 5014436 Mfr # G1077068
COMPARE
1/8 1/8 Straight
₹23,700.00INR / Each
PharmaFluor Panel-Mount Compression Fitting, Straight Union, PFA, 3/8" OD 5014437 Mfr # G1077014
COMPARE
3/8 3/8 Straight
₹18,900.00INR / Each