PharmaFluor® Compression to NPT(F) Threaded Fittings, Elbow Adapter, PFA
High-purity compression fittings designed for critical fluid and gas applications
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
16,600.00 - ₹21,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Flow Pattern Thread size NPT(F) (port 2) (in)
Availability Pricing
PharmaFluor Compression to Threaded Fitting, Elbow Adapter, PFA, 1/2" OD x 1/2" NPT(F) 5014419 Mfr # G1077096
COMPARE
1/2 Elbow 1/2
₹17,400.00INR / Each
PharmaFluor Compression to Threaded Fitting, Elbow Adapter, PFA, 1/2" OD x 3/8" NPT(F) 5014420 Mfr # G1077322
COMPARE
1/2 Elbow 3/8
₹21,400.00INR / Each
PharmaFluor Compression to Threaded Fitting, Elbow Adapter, PFA, 3/8" OD x 3/8" NPT(F) 5014421 Mfr # G1077095
COMPARE
3/8 Elbow 3/8
₹16,600.00INR / Each