pH Sensors
Nontoxic gel prevents silver/protein interference on the diaphragm
  • Pre-autoclaved
  • MSDS and GLP certificate available upon request
These pH sensors use a nontoxic gel to prevent silver/protein interference on the diaphragm. Each sensor has a Ti layer on the innertube to ensure no window between sensor head and gel which results in an excellent screen to protect against electromagnetic interference. Pre-autoclaved; MSDS and GLP certificate available upon request.
246,000.00 - ₹326,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Applikon Z001012051 pH Sensor for Fermentation Systems, 120 mm 2920720 Mfr # Z001012051
COMPARE
pH Sensor for Fermentation Systems, 120 mm
₹246,000.00INR / Each
Applikon Z001032551 pH Sensor for Fermentation System, 325 mm 2920724 Mfr # Z001032551
COMPARE
pH Sensor for Fermentation System, 325 mm
₹254,000.00INR / Each
Applikon Z001042551 pH Sensor for Fermentation System, 425 mm 2920726 Mfr # Z001042551
COMPARE
pH Sensor for Fermentation System, 425 mm
₹326,000.00INR / Each