Pesticides Cabinets

Reduce fire risks and protect workers
  • Store chemicals safely in the workplace
  • Enhance fire safety through door-lock system, and fireproof and explosion-proof vents
  • FM and CE certifications in strict compliance with FM approval standard for storage cabinets
Use to store insecticides, pesticides, disinfectors, and other chemicals. Suitable for lawn care, farms, parks, and other places.

Safety storage cabinets feature 18-gauge steel, all-welded, double fireproof steel panel, insulating air barrier
construction for protection and durability. Fireproofing vents at both sides of the cabinet (one on the top and
another on the bottom) effectively reduce the concentration of volatile substances and prevent fire hazards.
Reinforced shelves are adjustable and feature built-in troughs and leakproof wells to contain spills. The
manually-closing doors can be fully opened to 180° for easy access. Three-point linkage latch can be
padlocked to ensure complete closure for better security.
In storing chemicals, cabinet colors and warning labels are generally used to identify different chemicals, so if
a fire occurs, it is easy for firemen to quickly locate hazardous chemicals.
64,400.00 - ₹123,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Number of Shelves Description
Availability Pricing
SYSBEL WA810120G 12 Gal/45L,1 door, manual 1597692 Mfr # WA810120G
COMPARE
45 1 Pesticides Cabinets; 12 Gal/45L,1 door, manual
₹64,400.00INR / Each
SYSBEL WA810300G 30 Gal/114L,2 door, manual 1597693 Mfr # WA810300G
COMPARE
114 1 Pesticides Cabinets; 30 Gal/114L,2 door, manual
₹97,700.00INR / Each
SYSBEL WA810450G 45 Gal/170L,2 door, manual 1597694 Mfr # WA810450G
COMPARE
170 2 Pesticides Cabinets; 45 Gal/170L,2 door, manual
₹123,000.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat