Partlow 10in Circular Chart Recorders and Recording Controllers

Proven recording reliability that became the industry standard!
  • Capable of measuring, displaying, recording and controlling up to two process variables from a variety of inputs
  • All user entered data can be protected against unauthorized changes
  • Straightforward programming allows you to configure your recorder with a logical step-by-step process using a simple keypad
  • 4-digit display per pen
  • Universal Isolated sensor input: thermocouple, RTD, mA, mV, or V
  • Chart speed from 0.1 to 999.9 hours
  • Disposable fiber tipped pens: red, green
589,000.00 - ₹812,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chart Diameter (in) Chart Speed Number of Channels
Availability Pricing
Partlow 710000000021 10" Chart Recorder; 1 Channel WW-49001-88 Mfr # 710000000021
COMPARE
10 0.1 to 999.9 hours 1
₹589,000.00INR / Each
Partlow 710200000021 10" Chart Controller/Recorder; 1 Channel/2 Relay WW-49001-91 Mfr # 710200000021
COMPARE
10 0.1 to 999.9 hours 1
₹663,000.00INR / Each
Partlow 711200000021 10" Chart Controller/Recorder; 2 Channel/2 Relay WW-49001-92 Mfr # 711200000021
COMPARE
10 0.1 to 999.9 hours 2
₹812,000.00INR / Each
Partlow 711000000021 2 Chnl Chart Recorder Std Cover 90-115 Vac WW-49001-93 Mfr # 711000000021
COMPARE
10 0.1 to 999.9 hours 2
₹784,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat