Ovation Adjustable-Volume Pipettors
Proven ergonomically correct pipette is comfortable and easy to use!
  • Minimize contact pressure, tension, and exertion
  • Eliminate the need for organizers — the Ovation pipet stands on its own!
  • Flawless tipping and detipping, and simple menu make it simple to use
The Ovation design changes the concept of liquid transfer by reducing the causes of pain, fatigue, and stress from pipetting. Work for hours without tensions in the shoulder or elbow! Never over-rotate your wrist or hand — the Ovation pipettor was formulated to keep your hand in the neutral position recommended by ergonomic experts. A low-force, form-fitting plunger manually controls aspiration and dispensing speeds with minimal effort.
The Ovation pipettor is acceptable for use with especially fragile membranes that can easily be disrupted or fluids that tend to bubble. Handling is also fast and easy with the unique tip acquisition system that delivers an audible "click" to indicate that all tips have been securely fitted. Just the touch of a button immediately ejects all tips.
The large LCD display and motor-driven volume adjustments offer precise transfer, with up to five "one-button" presets. Set up and stored your most used settings for routine use and later recall. Other volume settings can be selected at precise increments throughout the pipettor's range. Pipettor accommodates both left- or right-handed users. Return to the factory calibration setting with the push of a button, making in-lab calibration simple.
115,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL)
Availability Pricing
VistaLab Ovation QS Quick Set Digital Ergonomic Single-Channel Pipette, 1 to 10 uL, Orange WW-06197-32 Mfr # 1057-0010
COMPARE
10 1
₹115,000.00INR / Each
VistaLab Ovation QS Quick Set Digital Ergonomic Single-Channel Pipette, 2 to 20 uL, Yellow WW-06197-34 Mfr # 1057-0020
COMPARE
20 2
₹115,000.00INR / Each
VistaLab Ovation QS Quick Set Digital Ergonomic Single-Channel Pipette, 10 to 100 uL, Green WW-06197-36 Mfr # 1057-0100
COMPARE
100 10
₹115,000.00INR / Each
VistaLab Ovation QS Quick Set Digital Ergonomic Single-Channel Pipette, 20 to 200 uL, Blue WW-06197-38 Mfr # 1057-0200
COMPARE
200 20
₹115,000.00INR / Each
VistaLab Ovation QS Quick Set Digital Ergonomic Single-Channel Pipette, 100 to 1000 uL, Purple WW-06197-40 Mfr # 1057-1000
COMPARE
1000 100
₹115,000.00INR / Each