Open Top Tank Systems
Open top tank systems provide maximum chemical handling options
Available in flat-bottom and total-drain conical bottom, HDLPE tanks are FDA compliant and meet NSF standard 61. Open-top tanks come equipped with a standard lid cover and molded-in gallon/liter graduation indicators. Lids have an all-plastic hinge and incorporate rib design to better support top-fitting installations. Lid assemblies enable access to tank interior without removing entire lid. Conical bottom tanks are designed to withstand weight of 1.9 specific gravity material in tanks. To accommodate floor plumbing and pumps, tank stands have a nominal clearance of 18” from bottom of tank to floor. Tanks do not come with stands, order separately.
111,000.00 - ₹780,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Volume (Gal) Volume (Liters)
Availability Pricing
Snyder 56800VOT45 Flat-Bottom Open Top Tank System, HDPE, 55 gallon 0696034 Mfr # 56800VOT45
COMPARE
HDLPE 55 208
₹111,000.00INR / Each
Snyder 56900VOT45 Flat-Bottom Open Top Tank System, HDPE, 90 gallon 0696036 Mfr # 56900VOT45
COMPARE
HDLPE 90 341
₹146,000.00INR / Each
Snyder 5780101N45 Open Top Tank, conical bottom, 500GAL 0696076 Mfr # 5780101N45
COMPARE
HDLPE 500 1893
₹780,000.00INR / Each