Ohaus® PR Series NTEP-Approved Toploading Balances
Reliable performance at an affordable price
  • Bright, easy-to-read backlit LCD
  • RS-232 interface allows connection to printer or PC
  • Small footprint saves desktop space
  • Selectable environmental filters compensate for harsh environments
  • Removable stainless steel pan for easy cleaning
  • NTEP Legal-For-Trade and Measurement Canada approved
The NTEP-certified toploading balances provide basic weighing, parts counting, and percent weighing applications. The backlit LCD and easy-to-use interface make these balances simple to operate. A standard RS-232 port provides communication with point-of-sale systems or printers.
The balances are designed with a durable ABS housing and a removable stainless steel pan. Other features include integrated weigh-below hook, security bracket, calibration lock, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, and stability indicator.
280,000.00 - ₹435,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus PR322N/E Toploading Balance, NTEP Approved; 320 g x 0.01 g 1180373 Mfr # PR322N/E
COMPARE
External 320 0.01
₹343,000.00INR / Each
Ohaus PR522N/E Toploading Balance, NTEP Approved; 520 g x 0.01 g 1180374 Mfr # PR522N/E
COMPARE
External 520 0.01
₹435,000.00INR / Each
Ohaus PR822N/E Toploading Balance, NTEP Approved; 820 g x 0.01 g 1180375 Mfr # PR822N/E
COMPARE
External 820 0.01
₹280,000.00INR / Each
Ohaus PR2202N/E Toploading Balance, NTEP Approved; 2,200 g x 0.01 g 1180376 Mfr # PR2202N/E
COMPARE
External 2200 0.01
₹312,000.00INR / Each
Ohaus PR5202N/E Toploading Balance, NTEP Approved; 5,200 g x 0.01 g 1180377 Mfr # PR5202N/E
COMPARE
External 5200 0.01
₹435,000.00INR / Each
Ohaus PR6201N/E Toploading Balance, NTEP Approved; 6,200 g x 0.1 g 1180380 Mfr # PR6201N/E
COMPARE
External 6200 0.1
₹349,000.00INR / Each