Ohaus Microplate Vortex Mixers
Designed for continuous uniform mixing of microplates
  • Safe to mix at high speeds
  • Variable speeds up to 3500 rpm
  • Effective speed control from low to high-speed mixing
  • Compact design takes up less benchtop space
  • 2 year warranty
The microplate vortex mixers are engineered for durability and efficiency, featuring a high-quality German motor designed for optimal mixing of microplates across a wide speed range. They are designed for uninterrupted use in the lab, ensuring reliability during extended mixing tasks.
A unique locking design ensures safe operation across the entire speed range, with microplate accessories securely in place throughout the entire speed range. A versatile speed control allows you to adjust from gentle shaking to vigorous vertexing, accommodating a wide range of sample types.
These mixers are perfect for laboratories needing reliable performance and flexibility in their mixing applications. The digital model provides precise speed control and a timer for consistent results, while non-digital model offers a cost-effective alternative with variable speed settings.
156,000.00 - ₹182,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Display Max Speed (rpm)
Availability Pricing
Ohaus Microplate Digital Vortex Mixer, 300 to 3500 rpm, 2 Microplate Capacity; 120 VAC 5302762 Mfr # VXMPDG
COMPARE
2 microplates Digital 3500
₹182,000.00INR / Each
Ohaus Microplate Analog Vortex Mixer, 300 to 3500 rpm, 2 Microplate Capacity; 120 VAC 5302763 Mfr # VXMPAL
COMPARE
2 microplates Analog 3500
₹156,000.00INR / Each