Ohaus® Defender® 3000 Bench Scale Bases
Economical yet durable weighing bases are ideal for industrial weighing
  • 304 stainless steel (SS) platform with stand rough environments
  • NTEP certified and Measurement Canada Class III certified
These heavy-duty scale bases feature removable 304 stainless steel platform for quick and easy cleaning. Mix and match with Defender 3000 indicators and mount system to create a scale that matches your application. All models feature single-point load cell. Select from bases with aluminum load cell that is sealed against moisture or washdown bases with silicon sealed IP67-rated stainless steel load cell.
204,000.00 - ₹548,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D15B1R Defender® 3000 Base, Aluminum Load Cell, 14"x12" Platform, 15kg x 2g 1134869 Mfr # D15B1R
COMPARE
15 14 12
₹204,000.00INR / Each
Ohaus D30B1R Defender® 3000 Base, Aluminum Load Cell, 14"x12" Platform, 30kg x 5g 1134870 Mfr # D30B1R
COMPARE
30 14 12
₹204,000.00INR / Each
Ohaus D75B1R Defender® 3000 Base, Aluminum Load Cell, 14"x12" Platform, 75kg x 10g 1134871 Mfr # D75B1R
COMPARE
75 14 12
₹204,000.00INR / Each
Ohaus D75B1L Defender® 3000 Base, Aluminum Load Cell, 21.7"x16.5" Platform, 75kg x 10g 1134872 Mfr # D75B1L
COMPARE
75 21 3/4 16 1/2
₹210,000.00INR / Each
Ohaus D150B1L Defender® 3000 Base, Aluminum Load Cell, 21.7"x16.5" Platform, 150kg x 20g 1134873 Mfr # D150B1L
COMPARE
150 21 3/4 16 1/2
₹210,000.00INR / Each
Ohaus D150B1X Defender® 3000 Base, Aluminum Load Cell, 25.6"x19.7" Platform, 150kg x 20g 1134874 Mfr # D150B1X
COMPARE
150 25 39/64 19 3/4
₹235,000.00INR / Each
Ohaus D300B1X Defender® 3000 Base, Aluminum Load Cell, 25.6"x19.7" Platform, 300kg x 50g 1134875 Mfr # D300B1X
COMPARE
300 25 39/64 19 3/4
₹235,000.00INR / Each
Ohaus D300B1V Defender® 3000 Base, Aluminum Load Cell, 31.5"x23.6" Platform, 300kg x 50g 1134876 Mfr # D300B1V
COMPARE
300 31 1/2 23 39/64
₹318,000.00INR / Each
Ohaus D600B1V Defender® 3000 Base, Aluminum Load Cell, 31.5"x23.6" Platform, 600kg x 100g 1134877 Mfr # D600B1V
COMPARE
600 31 1/2 23 39/64
₹318,000.00INR / Each
Ohaus D15C1R Defender® 3000 Base, IP67-Rated Load Cell, 14"x12" Platform, 15kg x 2g 1134878 Mfr # D15C1R
COMPARE
15 14 12
₹223,000.00INR / Each
Ohaus D30C1R Defender® 3000 Base, IP67-Rated Load Cell, 14"x12" Platform, 30kg x 5g 1134879 Mfr # D30C1R
COMPARE
30 14 12
₹260,000.00INR / Each
Ohaus D75C1R  Defender® 3000 Base, IP67-Rated Load Cell, 14"x12" Platform, 75kg x 10g 1134880 Mfr # D75C1R
COMPARE
75 14 12
₹260,000.00INR / Each
Ohaus D75C1L Defender® 3000 Base, IP67-Rated Load Cell, 21.7"x16.5" Platform, 75kg x 10g 1134881 Mfr # D75C1L
COMPARE
75 21 3/4 16 1/2
₹395,000.00INR / Each
Ohaus D150C1L Defender® 3000 Base, IP67-Rated Load Cell, 21.7"x16.5" Platform, 150kg x 20g 1134882 Mfr # D150C1L
COMPARE
150 21 3/4 16 1/2
₹395,000.00INR / Each
Ohaus D150C1X Defender® 3000 Base, IP67-Rated Load Cell, 25.6"x19.7" Platform, 150kg x 20g 1134883 Mfr # D150C1X
COMPARE
150 25 39/64 19 3/4
₹548,000.00INR / Each
Ohaus D300C1X Defender® 3000 Base, IP67-Rated Load Cell, 25.6"x19.7" Platform, 300kg x 50g 1134884 Mfr # D300C1X
COMPARE
300 25 39/64 19 3/4
₹548,000.00INR / Each
Showing 1- 16 of 16